Squalan 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215351000
215355000
215350025
ACRO215351000EA
567
DKK
ACRO215351000
ACRO215355000
ACRO215350025
Squalan 99%
Squalan
Formel:
C₃₀H₆₂ Kogepunkt: 210…215 °C (1 mmHg) Massefylde: 0,8085 g/cm³ Flammepunkt: 217 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008953 CAS-nummer: 111-01-3 EINECS: 203-825-6 Merck Index: 12,08923 |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4510 to 1.4530 (20°C, 589 nm) |
Density | (20°C) 0.807 to 0.810 g/ml |
GC | ≥98.5 % |
Iodine value | ≤1 g I/100 g |
Acid value | ≤0.2 mg KOH/g |
Saponification value | ≤1 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...