Nitrogenkolber, rund bund (Schlenk-beholdere)
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
03360122 03360128 03360137 03360437 03360449 03360458 03360470 03360428 03360149
201-1130EA 503 DKK
201-1130 201-1131 201-1132 201-1135 201-1136 201-1137 201-1138 LENZ03360428 LENZ03360149
Nitrogenkolber, rund bund (Schlenk-beholdere)
Kolber Nitrogenflasker
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With conical ground joint, NS stopcock and inscription label.