Stophaner til buretter, nåleventil
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
02700902 02702902
LENZ02700902EA 333 DKK
LENZ02700902 LENZ02702902
Stophaner til buretter, nåleventil
Stophaner
Needle-valve stopcocks, DURAN® housing with facetted tip, straight or lateral.
  • PTFE spindle
  • Cap and security device of PP
  • Valve bore of 0 to 2,5 mm