Stophaner, tovejs
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
02401402 02401804 02431402 02431804
227-3123EA 559 DKK
227-3123 LENZ02401804 LENZ02431402 LENZ02431804
Stophaner, tovejs
Stophaner
Stopcocks are made of DURAN® borosilicate glass with solid NS glass plug, screwthread retaining device, and two oblique bores.
  • Excellent chemical resistance
  • Excellent temperature resistance
With two oblique bores.
Leakage rate accoring to DIN 12540-1.