Slangekonnektorer, Y-formede
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
01600708 01600710
229-3532EA 39.9 DKK
229-3532 229-3533
Slangekonnektorer, Y-formede
Fittings og stik
Tubing connectors made of DURAN® borosilicate glass,Y-shaped with three glass olives.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • With tapered endpieces to ensure that tubes are held securely