Indsatsfiltre, Polydisc™ AS, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6724-5002 6724-5045 6724-5145
516-1108EA 2150 DKK
516-1108 516-1109 WHAT6724-5145
Indsatsfiltre, Polydisc™ AS, Whatman™
Filtre Indsatsfiltre
Whatman Polydisc AS (Aqueous Solution) in-line filters from Cytiva feature high-throughput PES membrane with a glass microfiber prefilter. Polydisc comes presterilized in medical-grade blister packs
  • Whatman filters with polyethersulfone membranes have low protein binding and are surfactant-free
  • GMF prefilter removes heavy particulates, increasing membrane longevity
  • Radiation sterilized, so there are no ethylene oxide (EtO) residuals
  • Barbed hose connections fit multiple tubing sizes
  • Integrity-testable with the bubble point method
  • Lightweight design prevents