Kemikaliebestandige overalls, 4530
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4530M 4530L 4530XL 4530XXL 4530S
113-0594EA 158 DKK
113-0594 113-0595 113-0596 113-0597 MMMA4530S
Kemikaliebestandige overalls, 4530
Engangsbeklædning
These disposable overalls are made of lightweight, breathable PP with flame resistance. Designed to be worn over primary FR garments, they provide protection against dusts and light liquid splashes.
  • Flame resistant treatment offers secondary protection against heat and flame
  • Breathable material helps to reduce heat build-up
  • Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures
  • Reinforcement gusset between the legs to help provide increased durability during bending and squatting
  • Two-way zip with sealable storm flap, three panel hood design
  • Elastic waist and ankles for convenience and freedom of movement
  • Anti-static coating on both sides helps reduce static build-up and prevent the risk of static discharge during use
Typical applications for this safety work wear may include: welding, metal grinding, general powder handling, and woodworking.
Colour: Blue (white panel on the back).
Certificeringer: Type 5/6, EN 1073-2, EN 1149-5, ISO 14116; Cat. III.