Membranfiltre, Nuclepore™ track etched, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10417006 155846 10417001 10417101 10417301 10417401 10417501 10417106 10417206 10417306 10417406 10417506 10417309 10417012 10417112 10417212 10417312 10417412 10417512 10417014 10417114 10417414 10417018 10417118 10417031 10417004 10417104 10417304 10418718 10418739 10418401 10418301 10418701 10418506 10418512 10418806 10418412 10418704 10418501 10418406 10418706 10418712 10418312 10418812 10418318 10418818 10418731 10418514 10417050 10417039 10419306 10419312 10419331 10419501 10419504 10419506 10419512 10419518 10419531 10419539 10419318 10419339
WHAT10417006EA 973 DKK
WHAT10417006 515-2292 WHAT10417001 WHAT10417101 WHAT10417301 WHAT10417401 WHAT10417501 WHAT10417106 WHAT10417206 WHAT10417306 WHAT10417406 WHAT10417506 WHAT10417309 WHAT10417012 WHAT10417112 WHAT10417212 514-1412 WHAT10417412 WHAT10417512 WHAT10417014 WHAT10417114 WHAT10417414 WHAT10417018 WHAT10417118 WHAT10417031 WHAT10417004 WHAT10417104 WHAT10417304 WHAT10418718 WHAT10418739 WHAT10418401 WHAT10418301 WHAT10418701 WHAT10418506 WHAT10418512 WHAT10418806 WHAT10418412 WHAT10418704 WHAT10418501 WHAT10418406 WHAT10418706 514-1413 514-1415 514-1414 WHAT10418318 WHAT10418818 WHAT10418731 WHAT10418514 WHAT10417050 WHAT10417039 WHAT10419306 WHAT10419312 WHAT10419331 WHAT10419501 WHAT10419504 WHAT10419506 WHAT10419512 WHAT10419518 WHAT10419531 WHAT10419539 WHAT10419318 WHAT10419339
Membranfiltre, Nuclepore™ track etched, Whatman™
Filtre Membranfiltre
Nuclepore™ track-etched hydrophilic membrane filters offer a highly defined pore size cut-off in the submicron range, enabling particle deformity measurement and capture of extremely small particles. The membranes have sharply defined pores, high flow rates, and excellent chemical and thermal resistance. They are suitable for a variety of samples and applications, including microscopy, cytology, parasitology, and certain air and oceanography analyses.
  • Track-etched for highly defined pore size and high pore density
  • Treated with polyvinylpyrrolidone (PVP) to render it hydrophilic
  • Low extractables, minimizing filtration sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat membrane surface for high particle visibility
Cytiva's Whatman Nuclepore hydrophilic membrane filter range is manufactured from high-quality polycarbonate film track-etched (PFTE) membrane for high nominal pore size precision.
These sharply defined cylindrical micropores, combined with high flow rates and chemical and thermal resistance, make Nuclepore filtration membranes suitable for a variety of samples and applications, including microscopy, cytology, parasitology, and certain air and oceanography analyses.
* PVP-free, hydrophobic.