Målekolber, trapezformede, klasse A, blå inddeling
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2960233 2960240 2960252 2960253 2960260 2960261 2960265 2960270 2960275
HIRS2960233EA 265 DKK
HIRS2960233 HIRS2960240 HIRS2960252 HIRS2960253 HIRS2960260 HIRS2960261 HIRS2960265 HIRS2960270 HIRS2960275
Målekolber, trapezformede, klasse A, blå inddeling
Kolber Volumetriske kolber
DURAN®, borosilicate glass, with hollow glas stopper, conformity certified (DE-M marking).
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck
Certificeringer: DIN EN ISO 1042