Silikonegummipakninger, septa
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
01341214 01341618 01342225 01342932 01344245
LENZ01341214EA 1230 DKK
LENZ01341214 LENZ01341618 LENZ01342225 LENZ01342932 LENZ01344245
Silikonegummipakninger, septa
Septa
Silicone, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Flexible
For piercing, for screw caps with bore.