3-(Brommethyl)heptan 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-2-ethylhexane, 2-Ethylhexylbromide, 3-(Bromomethyl)heptane

305331000 305335000
ACRO305331000EA 506 DKK
ACRO305331000 ACRO305335000
3-(Brommethyl)heptan 95%
2-Ethylhexyl bromide
Formel: C₈H₁₇Br
Kogepunkt: 75…77 °C (16 mmHg)
Massefylde: 1,08 g/cm³
Flammepunkt: 69 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000220
CAS-nummer: 18908-66-2
EINECS: 242-659-9

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4560 (20°C, 589 nm)
GC ≥94 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR