1-Brom-3-methylbutan ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Methylbutylbromide
A14492.0B
A14492.0I
A14492.30
A14492.0BEA
1530
DKK
A14492.0B
A14492.0I
A14492.30
1-Brom-3-methylbutan ≥98%
1-Bromo-3-methylbutane
Purity: 98%
CAS No.: 107-82-4
Molecular Formula: C5H11Br
Formula Weight: 151.05
Flash Point: 22°(71°F)
Refractive Index: 1.4409
Melting Point: -112°
Boiling Point: 120-121°
Beilstein Reference: 773652
Merck Reference: 14,5114
UN No.: 2341
MDL No.: MFCD00000253
Formel:
C₅H₁₁Br Molekylvægt: 151.05 g/mol Kogepunkt: 120…121 °C Smeltepunkt: –112 °C Massefylde: 1.208 Flammepunkt: 22 °C (71 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000253 CAS-nummer: 107-82-4 EINECS: 203-522-9 UN nummer: 2341 ADR: 3 III Merck Index: 13,05130 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...