n-Triacontan ≥99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Purity: 99%
CAS No.: 638-68-6
Molecular Formula: CH3(CH2)28CH3
Formula Weight: 422.83
Flash Point: 237°(458°F)
Odour: Odourless
Boiling Point: 455°
Beilstein Reference: 1777281
MDL No.: MFCD00009410
Formel:
H₃C(CH₂)₂₈CH₃ Kogepunkt: 455 °C Smeltepunkt: 65…68 °C Massefylde: 0,7797 g/cm³ (20 °C) Flammepunkt: 237 °C (458 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009410 CAS-nummer: 638-68-6 EINECS: 211-349-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...