n-Tetracontan 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

309165000 309160025
ACRO309165000EA 520 DKK
ACRO309165000 ACRO309160025
n-Tetracontan 98%
n-Tetracontane
Formel: H₃C(CH₂)₃₈CH₃
Molekylvægt: 563.09 g/mol
Smeltepunkt: 80…83 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015267
CAS-nummer: 4181-95-7
EINECS: 224-055-7

Bestil nu

Specifikationer Testresultater

Appearance White Powder or flakes
Infrared spectrum Conforms
Melting point 80 to 83 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR