Sikkerhedsbriller, 2800
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2800 2802 2805
111-1603EA 144 DKK
111-1603 MMMA2802 MMMA2805
Sikkerhedsbriller, 2800
Briller
These lightweight cover glasses (OTG) are designed to fit over most prescription spectacles with minimal interference. Available in three lens options. The green lens offers protection against infrared radiation arising from certain gas welding, gas cutting and brazing applications (IR shade 5).
  • Strong polycarbonate lens with anti-scratch coating
  • Adjustable temple length (four positions)
  • Inclinable lens for easy adjustment and maximum comfort
  • Soft low profile temples for comfort and minimum interaction with prescription spectacles
Weight: 38 g
Certificeringer: EN 166, EN 170, EN 171 (IR), FT1