2,2,3,3,4,4,5,5-Octafluor-1,6-hexandiol ≥97%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 1H,1H,6H,6H-Perfluoro-1,6-hexanediol, 1H,1H,6H,6H-Octafluoro-1,6-hexanediol
L16560.06 L16560.14
L16560.06EA 717 DKK
L16560.06 L16560.14
2,2,3,3,4,4,5,5-Octafluor-1,6-hexandiol ≥97%
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol
Purity: 97% CAS No.: 355-74-8 Molecular Formula: HOCH2(CF2)4CH2OH Formula Weight: 262.10 Melting Point: 67-70° Boiling Point: 100°/3mm MDL No.: MFCD00069087
Formel: C₆H₆F₈O₂
Kogepunkt: 100 °C (3 mmHg)
Smeltepunkt: 67…70 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00069087
CAS-nummer: 355-74-8