3-Phenyl-1-propyn ≥97% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Propynylbenzene

H53500.06 H53500.14
H53500.06EA 1810 DKK
H53500.06 H53500.14
3-Phenyl-1-propyn ≥97% stabilised
3-Phenyl-1-propyn

Stabilized


Purity: 97% CAS No.: 10147-11-2 Molecular Formula: C9H8 Formula Weight: 116.16 Flash Point: 52°(126°F) Refractive Index: 1.526 Boiling Point: 75°/20mm UN No.: 3295 MDL No.: MFCD00134431

Formel: C₉H₈
Kogepunkt: 75 °C (20 mmHg)
Massefylde: 0,934
Flammepunkt: 52 °C (126 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134431
CAS-nummer: 10147-11-2
UN nummer: 3295
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR