n-Octadecan ≥90%, tech.
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Octadecane
44301.A1
44301.A7
44301.A1EA
981
DKK
44301.A1
44301.A7
n-Octadecan ≥90%, tech.
n-Octadecane
Purity: 90%
CAS No.: 593-45-3
Molecular Formula: CH3(CH2)16CH3
Formula Weight: 254.50
Flash Point: 166°(330°F)
Physical Form: Crystalline mass/melt
Density: 0.782
Refractive Index: 1.4390
Appearance: White
Odour: Odourless
Melting Point: 28°
Boiling Point: 317°
MDL No.: MFCD00009007
Formel:
H₃C(CH₂)₁₆CH₃ Kogepunkt: 317 °C Smeltepunkt: 28 °C Massefylde: 0,782 Flammepunkt: 166 °C (330 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009007 CAS-nummer: 593-45-3 EINECS: 209-790-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...