Filter cartridges, Sartolon®
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5102507H1 5102507H2
SARP5102507H1EA 3660 DKK
SARP5102507H1 SARP5102507H2
Filter cartridges, Sartolon®
Filtre Filterpatroner
Sartolon® filter cartridges are designed for broad chemical compatibility for specific applications in the pharmaceutical and chemical industry. The double-layer design leads to superior filtration performance compared to other nylon membrane filters. Ideal for filtration of a broad range of solvents and solvent-containing liquids.
  • Withstand high differential pressures in either the forward or reverse direction of flow
Membrane: Hydrophilic nylon; support layers: PP; end cap, core and cage: PP; O-rings: silicone
Maximum differential pressure: fiveMore Product Information