Membranskivefiltre, Ultipor®
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NCG047100 NKG025100 NR013100 NR025100 NR047100 NR09025 NR10025 NR29325 NRG047100 NRG14225 NRG29325 NTG047100 NTG14225 NX013100 NX025100 NX047100 NX09025 NX14225 NX29325 NXG013100 NXG025100 NXG047100 NXG14225 NXG29325 NNG14225 NNG047100 NPG047100
516-9426EA 1710 DKK
516-9426 516-9428 516-9429 516-9430 516-9431 516-9433 516-9434 516-9436 516-9438 516-9442 516-9443 516-9447 516-9449 516-9451 516-9452 516-9453 516-9455 516-9457 516-9458 516-9459 516-9460 516-9461 516-9465 516-9466 PALLNNG14225 PALLNNG047100 PALLNPG047100
Membranskivefiltre, Ultipor®
Filtre Membranfiltre
Nylon 6,6 membranes for fluid filtration in the pharmaceutical, biotechnology, electronics, food and beverage industries and for analytical applications. These membranes are very resistant to mechanical, chemical and thermal stress. They can be used in either direction, are inherently hydrophilic, have high flow rates, low pressure drops and very low extractables. Where appropriate, they can be steam sterilised without change in flow or pore size characteristic.