Kompakt burette, med udskiftelig PTFE-stophane, brunt borosilikatglas, klasse B, SILBERBRAND
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13926 13928
BRND13926EA 978 DKK
BRND13926 BRND13928
Kompakt burette, med udskiftelig PTFE-stophane, brunt borosilikatglas, klasse B, SILBERBRAND
Buretter
Borosilicate glass 5.4, amber, class B.
  • Calibrated to deliver (TD, Ex)
  • White graduation
  • With reduced distance between subdivision marks for 25 ml burette
25 ml: Error limit according to class B, DIN EN ISO 385. 50 ml: DIN EN ISO 385, class B.
Leveringsinformation: Supplied with burette tube, stopcock with precision tip.