Kompakt burette, med udskiftelig PTFE-stophane, Schellbach-stribe, klasse AS, BLAUBRAND®
Leverandør: Brand
Ratings:
Gennemsntilig vurdering: 4.0 (Bedømmelse)
Samlet vurdering
Gennemsntilig vurdering
Var denne bedømmelse brugbar? Ja Nej
Ja: 2 Nej: 0 
Ja, jeg anbefaler
Très bon produit, je l'utilise tous les lundi en Tp pour réaliser des dosages. Cette burette permet un dosage très précis.
13916 13918 13913
612-7160EA 834 DKK
612-7160 612-7161 612-7159
Kompakt burette, med udskiftelig PTFE-stophane, Schellbach-stribe, klasse AS, BLAUBRAND®
Buretter
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Modular concept - all components, including PTFE stopcock, are replaceable
  • Schellbach stripe
Certificeringer: DIN EN ISO 385
Bestillingsoplysninger: Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.
Leveringsinformation: Includes burette tube with Schellbach stripe, stopcock with precision tip and batch certificate.