Acetone-dimethyl acetal 98+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
DMP
115630010
115635000
115630025
115630100
ACRO115630010EA
964
DKK
ACRO115630010
ACRO115635000
ACRO115630025
ACRO115630100
Acetone-dimethyl acetal 98+%
Acetone-dimethyl acetal
Formel:
C₅H₁₂O₂ Molekylvægt: 104.15 g/mol Kogepunkt: 83 °C (760 mmHg) Smeltepunkt: –47 °C Massefylde: 0.84 g/cm³ Flammepunkt: –5 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008479 CAS-nummer: 77-76-9 EINECS: 201-056-0 UN nummer: 3271 ADR: 3 II |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3770 to 1.3790 (20°C, 589 nm) |
Water | ≤0.5 % (K.F.) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...