Isopropylamin ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Propanamin

A15044.0F A15044.AE A15044.AP
A15044.0FEA 1270 DKK
A15044.0F A15044.AE A15044.AP
Isopropylamin ≥99%
Isopropylamin

Purity: 99+% CAS No.: 75-31-0 Molecular Formula: C3H9N Formula Weight: 59.11 Flash Point: -32°(-25°F) Physical Form: Clear liquid Density: 0.691 Refractive Index: 1.3750 Sensitivity: Air Sensitive Appearance: Colourless Odour: Amine-like Melting Point: -101° Boiling Point: 33-34° Beilstein Reference: 605259 Merck Reference: 14,5209 UN No.: 1221 MDL No.: MFCD00008082

Formel: C₃H₉N
Molekylvægt: 59.11 g/mol
Kogepunkt: 33…34 °C
Smeltepunkt: –101 °C
Massefylde: 0.691
Flammepunkt: –32 °C (–25 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008082
CAS-nummer: 75-31-0
EINECS: 200-860-9
UN nummer: 1221
ADR: 3 I
Merck Index: 13,05229

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR