6-Chlor-1-hexanol 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Chlorohexan-1-ol, 1-Chloro-6-hydroxyhexane, 6-Chlorohexanol
109280250
109281000
109285000
ACRO109280250EA
513
DKK
ACRO109280250
ACRO109281000
ACRO109285000
6-Chlor-1-hexanol 95%
6-Chloro-1-hexanol
Formel:
C₆H₁₃ClO Kogepunkt: 108…112 °C (14 mmHg) Massefylde: 1,03 g/cm³ Flammepunkt: 98 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002984 CAS-nummer: 2009-83-8 EINECS: 217-925-2 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4547 to 1.4567 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...