5-Chlor-1-pentanol ≥90.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C0815-25ML
C0815-5ML
TCIAC0815-25MLEA
895
DKK
TCIAC0815-25ML
TCIAC0815-5ML
5-Chlor-1-pentanol ≥90.0% (by GC)
5-Chloro-1-pentanol
Formel:
C₅H₁₁ClO Kogepunkt: 99°C Massefylde: 1,046 g/cm³ (17 °C) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00039556 CAS-nummer: 5259-98-3 EINECS: 226-067-8 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...