2-(2-Chlorethoxy)ethanol 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

172260500 172262500
ACRO172260500EA 426 DKK
ACRO172260500 ACRO172262500
2-(2-Chlorethoxy)ethanol 99%
2-(2-Chloroethoxy)ethanol
Formel: C₄H₉ClO₂
Kogepunkt: 79 °C (1013 hPa)
Massefylde: 1,18 g/cm³
Flammepunkt: 90 °C
MDL Number: MFCD00002870
CAS-nummer: 628-89-7
EINECS: 211-059-9
UN nummer: 3334
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4519 to 1.4539 (20°C, 589 nm)
Water ≤1 % (K.F.)
GC ≥98.5 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR