Chloroform ≥99.8% (by GC, corrected for water content) stabilised, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Trichlormethan

Stabilization: ~0.75% Stabilised with ethanol
Packaged under inert gas.

9257-03EA 2820 DKK
9257-03 9257-02
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Chloroform
Formel: CHCl₃
Molekylvægt: 119.38 g/mol
Kogepunkt: 61.7
Smeltepunkt: –63 °C
Massefylde: 1.4832 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000826
CAS-nummer: 67-66-3
EINECS: 200-663-8
UN nummer: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

For Laboratory,Research,or Manufacturing Use = 10
Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤10
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water) ≥99.8 %
Chloride (Cl) ≤10 ppm
Color (APHA) ≤10
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Titrable Acid (µeq/g) ≤0.5
Water (by KF, coulometric) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR