1,1,2,2-Tetrachlorethan 98.5%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

147940250 147941000 147940010 147940025
ACRO147940250EA 201 DKK
ACRO147940250 ACRO147941000 ACRO147940010 ACRO147940025
1,1,2,2-Tetrachlorethan 98.5%
1,1,2,2-Tetrachloroethane
Formel: Cl₂HCCHCl₂
Kogepunkt: 147 °C (760 mmHg)
Smeltepunkt: –43 °C
Massefylde: 1,58 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000848
CAS-nummer: 79-34-5
EINECS: 201-197-8
UN nummer: 1702
ADR: 6.1,II
Merck Index: 13,09264
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ad9914010b65d5d21f26f85ccb3fc00111fc8b3&vid=da3549d1364274a05dcc36a80f09745b568776d0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4950 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR