Bismuthtriacetat ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Acetic acid bismuth (III) salt, Bismuth (III) acetate, Bismuth(III) acetate
17574.18
17574.30
17574.18EA
1760
DKK
17574.18
17574.30
Bismuthtriacetat ≥99%
Bismuthtriacetat
Purity: 99%
CAS No.: 22306-37-2
Molecular Formula: C6H9BiO6
Formula Weight: 386.12
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Melting Point: dec.
MDL No.: MFCD00015629
Formel:
(H₃CCOO)₃Bi Molekylvægt: 386.11 g/mol Smeltepunkt: Dec. Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00015629 CAS-nummer: 22306-37-2 EINECS: 244-904-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...