Methylacetat, extra dry 99+%, AcroSeal®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Acetic acid methyl ester

371831000 371830010 371830025
371831000.EA 335 DKK
371831000. 371830010. 371830025.
Methylacetat, extra dry 99+%, AcroSeal®
Methylacetat
Formel: C₃H₆O₂
Molekylvægt: 74.08 g/mol
Kogepunkt: 57.4 °C (760 mmHg)
Smeltepunkt: –98 °C
Massefylde: 0.93 g/cm³
Flammepunkt: –10 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008711
CAS-nummer: 79-20-9
EINECS: 201-185-2
UN nummer: 1231
ADR: 3 II
Merck Index: 13,06038

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR