Ethylacetat, Prepsolv® for preparative chromatography, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Acetic acid ethyl ester
M-Clarity™ quality level = MQ200
1.13353.9030 1.13353.9190
1.13353.9030EA 0 DKK
1.13353.9030 1.13353.9190
Ethylacetat, Prepsolv® for preparative chromatography, Supelco®
Ethylacetat
Formel: CH₃COOC₂H₅
Molekylvægt: 88.11 g/mol
Kogepunkt: 77.1
Smeltepunkt: –83 °C
Massefylde: 0.902 g/cm³ (20 °C)
Flammepunkt: –4 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009171
CAS-nummer: 141-78-6
EINECS: 205-500-4
UN nummer: 1173
ADR: 3
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM