Ethylacetat ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
L10925.AK
L10925.AU
L10925.0F
L10925.AKEA
301
DKK
L10925.AK
L10925.AU
L10925.0F
Ethylacetat ≥99%
Ethylacetat
Purity: 99%
CAS No.: 141-78-6
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: -3°(26°F)
Refractive Index: 1.3720
Odour: Sweet
Melting Point: -84°
Boiling Point: 76-77°
Beilstein Reference: 506104
Merck Reference: 14,3757
UN No.: 1173
MDL No.: MFCD00009171
Formel:
CH₃COOC₂H₅ Kogepunkt: 76…77 °C Smeltepunkt: –84 °C Massefylde: 0,902 Flammepunkt: –3 °C (26 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009171 CAS-nummer: 141-78-6 EINECS: 205-500-4 UN nummer: 1173 ADR: 3,II REACH: 01-2119475103-46 Merck Index: 14,3757 |