Ethylacetat, extra dry 99.9%, AcroSeal®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Acetic acid ethyl ester

326901000 326900025 326900010
326901000.EA 538 DKK
326901000. 326900025. 326900010.
Ethylacetat, extra dry 99.9%, AcroSeal®
Ethylacetat
Formel: CH₃COOC₂H₅
Molekylvægt: 88.11 g/mol
Kogepunkt: 75…78 °C (760 mmHg)
Smeltepunkt: –83.5 °C
Massefylde: 0.902 g/cm³
Flammepunkt: –4 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009171
CAS-nummer: 141-78-6
EINECS: 205-500-4
UN nummer: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.85 %
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR