tert-Butylacetat 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,1-Dimethylethyl acetate

107765000 107760025
ACRO107765000EA 782 DKK
ACRO107765000 ACRO107760025
tert-Butylacetat 99%
tert-Butylacetat
Formel: C₆H₁₂O₂
Kogepunkt: 98 °C (760 mmHg)
Smeltepunkt: 90 - 97 °C
Massefylde: 0,86 g/cm³
Flammepunkt: 15 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008807
CAS-nummer: 540-88-5
EINECS: 208-760-7
UN nummer: 1123
ADR: 3,II
Merck Index: 13,01536

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3880 (20°C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.002 %
Titration with NaOH ≥88.0 %
Dilution test passes test
Chloride (Cl) ≤10 ppm
Sulfite (SO3) passes test
Sulfate (SO4) ≤20 ppm
Ammonium (NH4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Free acid (CH3COOH) ≤0.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR