Diacetyl ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,3-Butanedione

A14217.09 A14217.22 A14217.36
A14217.09EA 398 DKK
A14217.09 A14217.22 A14217.36
Diacetyl ≥99%
Diacetyl

Purity: 99% CAS No.: 431-03-8 Molecular Formula: C4H6O2 Formula Weight: 86.09 Flash Point: 10°(50°F) Refractive Index: 1.3940 Odour: Sweet Melting Point: -3° Boiling Point: 87-88° Beilstein Reference: 605398 Merck Reference: 14,2966 UN No.: 2346 MDL No.: MFCD00008756

Formel: C₄H₆O₂
Kogepunkt: 87…88 °C
Smeltepunkt: –3 °C
Massefylde: 0,983
Flammepunkt: 10 °C (50 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008756
CAS-nummer: 431-03-8
EINECS: 207-069-8
UN nummer: 2346
ADR: 3,II
Merck Index: 14,2966

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR