2,4-Pentandion ≥99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
ACAC, 2,4-Pentanedione
Purity: 99%
CAS No.: 123-54-6
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 34°(93°F)
Physical Form: Clear liquid
Density: 0.973
Refractive Index: 1.4520
Appearance: Colourless
Melting Point: -23°
Boiling Point: 140-141°
Beilstein Reference: 741937
Merck Reference: 14,81
UN No.: 2310
MDL No.: MFCD00008787
Formel:
CH₃COCH₂COCH₃ Kogepunkt: 140…141 °C Smeltepunkt: –23 °C Massefylde: 0,973 Flammepunkt: 34 °C (93 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008787 CAS-nummer: 123-54-6 EINECS: 204-634-0 UN nummer: 2310 ADR: 3,III Merck Index: 14,81 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...