Spande med snap-on låg
Leverandør: JOKEY PLASTIK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
101438/101425 102604/102842 101366/101570 105021/102842
216-0792EA 1090 DKK
216-0792 216-0793 216-0791 216-0794
Spande med snap-on låg
Spande
PP, opaque white.
  • Resistant to chemical products
  • Stackable and sturdy with plastic carrying handle
  • Uncapped, aseptic
Recyclable, single-use buckets for transport and deliveries; improves working conditions by eliminating washing and decanting tasks. Tamper-evident, skirted lid.