Volumetric flasks, class A, blue markings, BLAUBRAND® PURprotect
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36549 36548 36550 36551 36552 36553
612-3644EA 801 DKK
612-3644 612-3643 612-3645 612-3647 612-3648 612-3649
Volumetric flasks, class A, blue markings, BLAUBRAND® PURprotect
Kolber Volumetriske kolber
Borosilicate 3.3 glass, PUR plastic coated, with PP stopper.
  • Calibrated to contain (TC, In), incl. one batch certificate
  • Maximum operating temperature at dry heat is 135 °C (exposure time ˂30 min)
  • Frequent autoclaving at 121 °C reduces splintering protection
  • Cleaning up to max. 95 °C
  • DE-M marking
The PUR coating envelops the volumetric flasks like a protection skin. In the event of breakage, the safety coating helps contain the glass fragments. Compared to uncoated glass flasks the electrostatic charge is not increased. The light blue coating facilitates optical distinction.
*Bred hals.
Certificeringer: DIN EN ISO 1042.