Tragtstativ
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
74511 74506
BRND74511EA 2260 DKK
BRND74511 519-2711
Tragtstativ
Retortstativer
PP plates for holding two or four funnels with top outer-Ø of 50 to 120 mm.
  • With aluminum support rod (Ø×L: 12,7×595 mm)
  • Adjustable funnel height