PCR-varmeforseglingsfilm og -folier
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030127.838 0030127.854 0030127.781 0030127.790 0030132.947
613-3768EA 1560 DKK
613-3768 613-3769 713-0005 713-0006 713-0019
PCR-varmeforseglingsfilm og -folier
Forsegling Mikrotiterpladetætninger
Ensure airtight sealing of your PCR plates with Eppendorf® PCR films and foils. Tailor your choice to your assay needs, like the Masterclear® real-time PCR film, optimal for qPCR with its superior optical characteristics, or heat-sealing options for the best evaportion protection.
  • PCR plate seals offer excellent chemical resistance and have been tested for resistance to a variety of common laboratory substances
  • Specialised seals with optimal optical properties are available to help minimise signal loss in qPCR
Eppendorf PCR films and foils provide effective and airtight sealing for plates to help protect PCR samples during thermal cycling and storage.