3-Chlor-4-hydroxybenzaldehyd 98

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A17515.14 A17515.06
A17515.14EA 4160 DKK
A17515.14 A17515.06
3-Chlor-4-hydroxybenzaldehyd 98
3-Chloro-4-hydroxybenzaldehyde

Purity: 98% CAS No.: 2420-16-8 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Melting Point: 128-132° MDL No.: MFCD00016981

Formel: C₇H₅ClO₂
Molekylvægt: 156.57 g/mol
Kogepunkt: 251
Smeltepunkt: 128…132 °C
Massefylde: 1.0404 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016981
CAS-nummer: 2420-16-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR