3-Heptanon 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethylbutylketon
120380010
120382500
120380025
ACRO120380010EA
2290
DKK
ACRO120380010
ACRO120382500
ACRO120380025
3-Heptanon 98%
3-Heptanone
Formel:
C₇H₁₄O Kogepunkt: 146…149 °C (760 mmHg) Smeltepunkt: –39 °C Massefylde: 0,81 g/cm³ Flammepunkt: 41 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009483 CAS-nummer: 106-35-4 EINECS: 203-388-1 UN nummer: 1224 ADR: 3,III |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4075 to 1.4095 (20°C, 589 nm) |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...