Palladium(II) trifluoracetat 97%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Palladium(II) 2,2,2-trifluoroacetate
317020010 317020050
ACRO317020010EA 1120 DKK
ACRO317020010 ACRO317020050
Palladium(II) trifluoracetat 97%
Palladium(II) trifluoracetat
Formel: C₄F₆O₄Pd
Molekylvægt: 332.45 g/mol
Smeltepunkt: 210 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013204
CAS-nummer: 42196-31-6
Bestil nu
Specifikationer Testresultater
Appearance Tan to brown Powder or crystals
Infrared spectrum Conforms
Assay 30.9 to 33.2 % (Pd)