Natriumdodecylsulfat (SDS), LiChropur™ for ion pair chromatography, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
M-Clarity™ quality level = MQ100
1.18309.0025 1.18309.0100
1.18309.0025EA 4630 DKK
1.18309.0025 1.18309.0100
Natriumdodecylsulfat (SDS), LiChropur™ for ion pair chromatography, Supelco®
Natriumdodecylsulfat (SDS)
Formel: CH₃(CH₂)₁₁OSO₃Na
Smeltepunkt: 204…207 °C
Massefylde: 1,1 g/cm³ (20 °C)
Flammepunkt: >100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036175
CAS-nummer: 151-21-3
EINECS: 205-788-1
UN nummer: 1325
ADR: 4.1,
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM