Natriummetaborat tetrahydrat, tech.
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
36736.A1
36736.A7
36736.A1EA
783
DKK
36736.A1
36736.A7
Natriummetaborat tetrahydrat, tech.
Natriummetaborat tetrahydrat
CAS No.: 10555-76-7
Molecular Formula: NaBO2.4H2O
Formula Weight: 137.86 (65.80anhy)
Physical Form: -8 Mesh Crystalline
Melting Point: ca 53° dec.
MDL No.: MFCD00149205
Formel:
BNaO₂·4H₂O Molekylvægt: 137.86 g/mol Smeltepunkt: ∼ 53 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149205 CAS-nummer: 10555-76-7 EINECS: 231-891-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...