Ethynylcyclopentan ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H53483.03
H53483.06
H53483.14
H53483.03EA
654
DKK
H53483.03
H53483.06
H53483.14
Ethynylcyclopentan ≥97%
Ethynylcyclopentan
Purity: 97%
CAS No.: 930-51-8
Molecular Formula: C7H10
Formula Weight: 94.16
Flash Point: 1°(34°F)
Refractive Index: 1.432
Boiling Point: 107-109°
UN No.: 3295
MDL No.: MFCD00013744
Formel:
C₇H₁₀ Molekylvægt: 94.16 g/mol Kogepunkt: 107…109 °C Massefylde: 0.812 Flammepunkt: 1 °C (34 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013744 CAS-nummer: 930-51-8 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...