Benzamidin, HCl

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Benzimidamide hydrochloride, Benzamidinium chloride, Phenylamidine hydrochloride, Benzene-1-carboximidamide hydrochloride

14525.01 14525.02
SERA14525.01EA 0 DKK
SERA14525.01 SERA14525.02
Benzamidin, HCl
Benzamidin, HCl
Formel: C₇H₉ClN₂
Molekylvægt: 156.61 g/mol
Smeltepunkt: 174…179 °C
MDL Number: MFCD00066285
CAS-nummer: 1670-14-0
EINECS: 216-795-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR