Benzamidin, HCl, Calbiochem®
Leverandør: Merck Millipore Calbiochem
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Benzimidamide hydrochloride, Benzamidinium chloride, Phenylamidine hydrochloride, Benzene-1-carboximidamide hydrochloride
199001-25
199001-5
199001-25EA
1510
DKK
199001-25
199001-5
Benzamidin, HCl, Calbiochem®
Benzamidin, HCl
Formel:
C₇H₉ClN₂ Molekylvægt: 156.61 g/mol Smeltepunkt: 174…179 °C |
MDL Number:
MFCD00066285 CAS-nummer: 1670-14-0 EINECS: 216-795-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...