9,10-Anthraquinon, Supelco®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Anthracene-9,10-dione, Anthraquinone
M-Clarity™ quality level = MQ100
67372-1G
67372-250MG
67372-1GEA
1190
DKK
67372-1G
67372-250MG
9,10-Anthraquinon, Supelco®
9,10-Anthraquinon
Formel:
C₁₄H₈O₂ Molekylvægt: 208.22 g/mol Kogepunkt: 380 Smeltepunkt: 165…169 °C Massefylde: 1.419…1.438 g/cm³ (20 °C) Flammepunkt: 185 |
MDL Number:
MFCD00001188 CAS-nummer: 84-65-1 EINECS: 201-549-0 Merck Index: 13,00692 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...